siRNA Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191978.1 | NP_001178907.1 | 3 | 414 | ||
| XM_006237088.2 | XP_006237150.1 | 3 | 1990 | |||
| XM_006237089.2 | XP_006237151.1 | 2 | 735 | |||
| XM_006237090.2 | XP_006237152.1 | 2 | 1892 | |||
| GenBank mRNA | FQ227184.1 | - | 3 | 483 | ||
| FQ227324.1 | - | 3 | 483 | |||
| GU357482.1 | - | 3 | 264 | |||

Clec1b
Genomic Map