siRNA Details
| Gene Symbol: Bcl2l14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024338.1 | NP_001019509.1 | 4, 5 | 835 | ||
| XM_006237533.2 | XP_006237595.1 | 5, 6 | 981 | |||
| XM_006237534.2 | XP_006237596.1 | 3, 4 | 840 | |||
| XM_008763416.1 | XP_008761638.1 | 3, 4 | 665 | |||
| GenBank mRNA | BC079010.1 | - | 4, 5 | 835 | 7113223 | |

Bcl2l14
Genomic Map