siRNA Details
Gene Symbol: LOC500350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047957.1 | NP_001041422.1 | 6 | 965 | ||
GenBank mRNA | AY539890.1 | - | 6 | 965 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC500350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047957.1 | NP_001041422.1 | 6 | 965 | ||
GenBank mRNA | AY539890.1 | - | 6 | 965 |