siRNA Details
| Gene Symbol: ADIPOQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177800.1 | NP_001171271.1 | 4 | 495 | ||
| NM_004797.3 | NP_004788.1 | 3 | 444 | |||
| GenBank mRNA | AK291525.1 | - | 3 | 417 | ||
| AK312868.1 | - | 4 | 465 | |||
| AX767959.1 | - | 3 | 386 | |||
| BC054496.1 | - | 385 | 6192794 | |||
| BC096308.3 | - | 3 | 428 | 40007540 | ||
| BC096309.3 | - | 3 | 428 | 40007541 | ||
| BC096310.3 | - | 3 | 428 | 40007542 | ||
| BC096311.3 | - | 3 | 428 | 40007544 | ||
| D45371.1 | - | 3 | 386 | |||
| EU420013.1 | - | 2 | 360 | |||

ADIPOQ
Genomic Map