siRNA Details
| Gene Symbol: Tuba8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024339.1 | NP_001019510.1 | 4 | 911 | ||
| XM_008763254.1 | XP_008761476.1 | 4 | 1048 | |||
| GenBank mRNA | BC079185.1 | - | 4 | 911 | 7131106 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tuba8| Gene Symbol: Tuba8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024339.1 | NP_001019510.1 | 4 | 911 | ||
| XM_008763254.1 | XP_008761476.1 | 4 | 1048 | |||
| GenBank mRNA | BC079185.1 | - | 4 | 911 | 7131106 | |