siRNA Details
| Gene Symbol: RGD1561849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109260.1 | NP_001102730.1 | 3, 4 | 653 | ||
| GenBank mRNA | FQ211566.1 | - | 3, 4 | 567 | ||
| FQ212089.1 | - | 3, 4 | 577 | |||
| FQ212229.1 | - | 3, 4 | 577 | |||

RGD1561849
Genomic Map