siRNA Details
| Gene Symbol: Esrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127564.2 | NP_001121036.2 | 10 | 1453 | ||
| XM_006237876.2 | XP_006237938.1 | 10 | 1533 | |||
| XM_006237877.2 | XP_006237939.1 | 10 | 1533 | |||
| XM_006237878.2 | XP_006237940.1 | 10 | 1533 | |||
| XM_006237879.2 | XP_006237941.1 | 10 | 1532 | |||
| XM_006237880.2 | XP_006237942.1 | 10 | 1532 | |||
| GenBank mRNA | BC166735.1 | - | 10 | 1453 | 7936665 | |

Esrp1
Genomic Map