siRNA Details
| Gene Symbol: Esrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237876.2 | XP_006237938.1 | 14, 15 | 2320 | ||
| XM_006237877.2 | XP_006237939.1 | 14, 15 | 2308 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_194055, AK140848.

Esrp1
Genomic Map