siRNA Details
| Gene Symbol: Rmdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031663.1 | NP_001026833.1 | 5, 6 | 588 | ||
| GenBank mRNA | BC079473.1 | - | 5, 6 | 594 | ||
| BC098713.1 | - | 5, 6 | 588 | 7322407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rmdn1| Gene Symbol: Rmdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031663.1 | NP_001026833.1 | 5, 6 | 588 | ||
| GenBank mRNA | BC079473.1 | - | 5, 6 | 594 | ||
| BC098713.1 | - | 5, 6 | 588 | 7322407 | ||