siRNA Details
| Gene Symbol: Tstd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575783.6 | XP_575783.3 | 3 | 454 | ||
| GenBank mRNA | BC078951.1 | - | 3 | 422 | ||
| BC089864.1 | - | 3 | 434 | |||
| BC104709.1 | - | 3 | 337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tstd3| Gene Symbol: Tstd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575783.6 | XP_575783.3 | 3 | 454 | ||
| GenBank mRNA | BC078951.1 | - | 3 | 422 | ||
| BC089864.1 | - | 3 | 434 | |||
| BC104709.1 | - | 3 | 337 | |||