siRNA Details
| Gene Symbol: Spaca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191982.1 | NP_001178911.1 | 2, 3 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spaca1| Gene Symbol: Spaca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191982.1 | NP_001178911.1 | 2, 3 | 284 | ||