siRNA Details
| Gene Symbol: LOC100912292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282336.1 | NP_001269265.1 | 2 | 925 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100912292| Gene Symbol: LOC100912292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282336.1 | NP_001269265.1 | 2 | 925 | ||