siRNA Details
| Gene Symbol: LOC500445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024343.2 | NP_001019514.2 | 5 | 873 | ||
| XM_006238096.2 | XP_006238158.1 | 5 | 1444 | |||
| GenBank mRNA | BC082005.1 | - | 5 | 854 | 7131603 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500445| Gene Symbol: LOC500445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024343.2 | NP_001019514.2 | 5 | 873 | ||
| XM_006238096.2 | XP_006238158.1 | 5 | 1444 | |||
| GenBank mRNA | BC082005.1 | - | 5 | 854 | 7131603 | |