siRNA Details
| Gene Symbol: RGD1560723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763685.1 | XP_008761907.1 | 1 | 2563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560723| Gene Symbol: RGD1560723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763685.1 | XP_008761907.1 | 1 | 2563 | ||