siRNA Details
| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024344.1 | NP_001019515.1 | 5 | 517 | ||
| GenBank mRNA | BC088199.1 | - | 5 | 517 | 7377221 | |
| FQ225852.1 | - | 4 | 432 | |||
| FQ233921.1 | - | 4 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sit1| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024344.1 | NP_001019515.1 | 5 | 517 | ||
| GenBank mRNA | BC088199.1 | - | 5 | 517 | 7377221 | |
| FQ225852.1 | - | 4 | 432 | |||
| FQ233921.1 | - | 4 | 432 | |||