siRNA Details
| Gene Symbol: Chp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024139.2 | NP_077053.1 | 6, 7 | 664 | ||
| GenBank mRNA | AB070350.1 | - | 6, 7 | 649 | ||
| BC062029.1 | - | 6, 7 | 664 | 5623922 | ||
| U39875.1 | - | 6, 7 | 910 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chp1| Gene Symbol: Chp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024139.2 | NP_077053.1 | 6, 7 | 664 | ||
| GenBank mRNA | AB070350.1 | - | 6, 7 | 649 | ||
| BC062029.1 | - | 6, 7 | 664 | 5623922 | ||
| U39875.1 | - | 6, 7 | 910 | |||