siRNA Details
| Gene Symbol: Epb41l4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127565.1 | NP_001121037.1 | 7 | 702 | ||
| GenBank mRNA | BC166749.1 | - | 7 | 702 | 5599589 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epb41l4b| Gene Symbol: Epb41l4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127565.1 | NP_001121037.1 | 7 | 702 | ||
| GenBank mRNA | BC166749.1 | - | 7 | 702 | 5599589 | |