siRNA Details
| Gene Symbol: Rnf183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238243.2 | XP_006238305.1 | 6 | 1693 | ||
| XM_008763806.1 | XP_008762028.1 | 5 | 1808 | |||
| XM_008763807.1 | XP_008762029.1 | 3 | 986 | |||
| XM_008763808.1 | XP_008762030.1 | 5 | 1471 | |||
| XM_008763809.1 | XP_008762031.1 | 4 | 1298 | |||
| XM_008763810.1 | XP_008762032.1 | 2 | 798 | |||

Rnf183
Genomic Map