siRNA Details
| Gene Symbol: RGD1564074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763964.1 | XP_008762186.1 | 22 | 2455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564074| Gene Symbol: RGD1564074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763964.1 | XP_008762186.1 | 22 | 2455 | ||