siRNA Details
| Gene Symbol: TM9SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004800.2 | NP_004791.1 | 3, 4 | 566 | ||
| GenBank mRNA | AK093507.1 | - | 3, 4 | 265 | ||
| AK290514.1 | - | 3, 4 | 521 | |||
| AK315210.1 | - | 3, 4 | 521 | |||
| BC110656.1 | - | 3, 4 | 523 | 3855486 | ||
| BC130293.1 | - | 3, 4 | 400 | 40146324 | ||
| U81006.1 | - | 3, 4 | 466 | |||

TM9SF2
Genomic Map