siRNA Details
| Gene Symbol: Ybx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031563.3 | NP_113751.3 | 6 | 820 | ||
| XM_008764058.1 | XP_008762280.1 | 6 | 860 | |||
| XM_008764059.1 | XP_008762281.1 | 5 | 970 | |||
| GenBank mRNA | AB046928.2 | - | 6 | 979 | ||
| BC072486.1 | - | 6 | 820 | 7105037 | ||
| BC098672.1 | - | 6 | 820 | 7124218 | ||
| BC103637.1 | - | 6 | 814 | 7107271 | ||
| D13309.1 | - | 6 | 790 | |||
| FQ232156.1 | - | 3 | 462 | |||
| M57299.1 | - | 6 | 796 | |||
| M69138.1 | - | 6 | 956 | |||

Ybx1
Genomic Map