siRNA Details
| Gene Symbol: Cdca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025050.1 | NP_001020221.1 | 4, 5 | 467 | ||
| XM_008764064.1 | XP_008762286.1 | 5, 6 | 467 | |||
| GenBank mRNA | BC079274.1 | - | 4, 5 | 467 | 7134027 | |
| BC079293.1 | - | 5, 6 | 446 | 7134482 | ||

Cdca8
Genomic Map