siRNA Details
| Gene Symbol: RGD1560436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109264.1 | NP_001102734.1 | 3, 4 | 809 | ||
| XM_006238898.2 | XP_006238960.1 | 3, 4 | 856 | |||
| XM_006238900.2 | XP_006238962.1 | 4, 5 | 757 | |||
| XM_008764065.1 | XP_008762287.1 | 3, 4 | 859 | |||

RGD1560436
Genomic Map