siRNA Details
| Gene Symbol: Gmeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109268.2 | NP_001102738.1 | 1 | 159 | ||
| XM_008764165.1 | XP_008762387.1 | 1 | 492 | |||
| XM_008764166.1 | XP_008762388.1 | 2 | 855 | |||
| XM_008764167.1 | XP_008762389.1 | 2 | 852 | |||
| XM_008764168.1 | XP_008762390.1 | 3 | 743 | |||
| XM_008764169.1 | XP_008762391.1 | 2 | 686 | |||
| GenBank mRNA | BC158640.1 | - | 1 | 159 | 8362091 | |

Gmeb1
Genomic Map