siRNA Details
| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025772.1 | NP_001020943.1 | 5 | 558 | ||
| XM_008764278.1 | XP_008762500.1 | 5 | 816 | |||
| XM_008764279.1 | XP_008762501.1 | 5 | 463 | |||
| XM_008764280.1 | XP_008762502.1 | 4 | 714 | |||
| GenBank mRNA | BC083648.1 | - | 4 | 500 | ||
| BC098937.1 | - | 5 | 558 | 7453388 | ||

Stpg1
Genomic Map