siRNA Details
| Gene Symbol: Tmco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034949.1 | NP_001030121.1 | 13 | 1592 | ||
| XM_006239262.1 | XP_006239324.1 | 13 | 3017 | |||
| GenBank mRNA | BC099755.1 | - | 13 | 1592 | 5623720 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmco4| Gene Symbol: Tmco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034949.1 | NP_001030121.1 | 13 | 1592 | ||
| XM_006239262.1 | XP_006239324.1 | 13 | 3017 | |||
| GenBank mRNA | BC099755.1 | - | 13 | 1592 | 5623720 | |