siRNA Details
| Gene Symbol: Pax7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191984.1 | NP_001178913.1 | 2 | 167 | ||
| XM_006239263.2 | XP_006239325.1 | 2 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pax7| Gene Symbol: Pax7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191984.1 | NP_001178913.1 | 2 | 167 | ||
| XM_006239263.2 | XP_006239325.1 | 2 | 251 | |||