siRNA Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114599.2 | NP_001108071.1 | 4 | 1097 | ||
| XM_006239264.2 | XP_006239326.1 | 4 | 1125 | |||
| GenBank mRNA | BC083784.1 | - | 5 | 1179 | ||
| FQ222061.1 | - | 3 | 1032 | |||

Szrd1
Genomic Map