siRNA Details
| Gene Symbol: Fhad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750075.3 | XP_003750123.1 | 22, 23 | 2966 | ||
| XM_006239329.2 | XP_006239391.1 | 22, 23 | 2966 | |||
| XM_006239330.2 | XP_006239392.1 | 21, 22 | 2858 | |||
| GenBank mRNA | BC098024.1 | - | 12, 13 | 1490 | 7455859 | |

Fhad1
Genomic Map