siRNA Details
| Gene Symbol: Fhad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750075.3 | XP_003750123.1 | 20 | 2640 | ||
| XM_006239329.2 | XP_006239391.1 | 20 | 2640 | |||
| XM_006239330.2 | XP_006239392.1 | 19 | 2532 | |||
| GenBank mRNA | BC098024.1 | - | 10 | 1164 | 7455859 | |

Fhad1
Genomic Map