siRNA Details
| Gene Symbol: Tmem51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109273.1 | NP_001102743.1 | 3 | 1137 | ||
| XM_006239313.2 | XP_006239375.1 | 2 | 737 | |||
| XM_006239314.2 | XP_006239376.1 | 3 | 1478 | |||
| XM_006239315.2 | XP_006239377.1 | 3 | 973 | |||
| XM_006239317.2 | XP_006239379.1 | 3 | 2472 | |||
| XM_008764281.1 | XP_008762503.1 | 3 | 1202 | |||

Tmem51
Genomic Map