siRNA Details
| Gene Symbol: OTOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287489.1 | NP_001274418.1 | 35 | 4383 | ||
| NM_004802.3 | NP_004793.2 | 18 | 2181 | |||
| NM_194248.2 | NP_919224.1 | 35 | 4383 | |||
| NM_194322.2 | NP_919303.1 | 17 | 2335 | |||
| NM_194323.2 | NP_919304.1 | 18 | 2181 | |||
| XM_017005338.1 | XP_016860827.1 | 35 | 4196 | |||
| GenBank mRNA | AF107403.1 | - | 18 | 2181 | ||
| AF183185.1 | - | 35 | 4383 | |||
| AF183186.1 | - | 17 | 2335 | |||
| AF183187.1 | - | 18 | 2181 | |||
| AK296272.1 | - | 8 | 738 | |||

OTOF
Genomic Map