siRNA Details
| Gene Symbol: Atad2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025151.1 | NP_001020322.1 | 6 | 647 | ||
| GenBank mRNA | BC081936.1 | - | 6 | 647 | 7115192 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atad2b| Gene Symbol: Atad2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025151.1 | NP_001020322.1 | 6 | 647 | ||
| GenBank mRNA | BC081936.1 | - | 6 | 647 | 7115192 | |