siRNA Details
| Gene Symbol: Rnf144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082410.1 | NP_001075879.1 | 9 | 1109 | ||
| XM_008764604.1 | XP_008762826.1 | 8 | 1343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf144a| Gene Symbol: Rnf144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082410.1 | NP_001075879.1 | 9 | 1109 | ||
| XM_008764604.1 | XP_008762826.1 | 8 | 1343 | |||