siRNA Details
| Gene Symbol: Dcdc2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195807.1 | NP_001182736.1 | 1 | 141 | ||
| XM_006239955.2 | XP_006240017.1 | 1 | 512 | |||
| XM_006239956.2 | XP_006240018.1 | 1 | 511 | |||
| XM_006239957.2 | XP_006240019.1 | 1 | 511 | |||
| XM_006239958.2 | XP_006240020.1 | 1 | 511 | |||
| GenBank mRNA | CK596582.1 | - | 1 | 141 | ||

Dcdc2c
Genomic Map