siRNA Details
| Gene Symbol: LOC500684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047959.1 | NP_001041424.1 | 1 | 820 | ||
| GenBank mRNA | AY325221.1 | - | 1 | 820 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500684| Gene Symbol: LOC500684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047959.1 | NP_001041424.1 | 1 | 820 | ||
| GenBank mRNA | AY325221.1 | - | 1 | 820 | ||