siRNA Details
Gene Symbol: LOC500684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047959.1 | NP_001041424.1 | 1 | 470 | ||
GenBank mRNA | AY325221.1 | - | 1 | 470 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC500684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047959.1 | NP_001041424.1 | 1 | 470 | ||
GenBank mRNA | AY325221.1 | - | 1 | 470 |