siRNA Details
| Gene Symbol: Plekhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127566.1 | NP_001121038.1 | 8 | 921 | ||
| GenBank mRNA | BC161896.1 | - | 8 | 921 | 7319876 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhd1| Gene Symbol: Plekhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127566.1 | NP_001121038.1 | 8 | 921 | ||
| GenBank mRNA | BC161896.1 | - | 8 | 921 | 7319876 | |