siRNA Details
| Gene Symbol: Map3k9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100872.1 | NP_001094342.1 | 2 | 776 | ||
| XM_006240303.2 | XP_006240365.1 | 2 | 999 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k9| Gene Symbol: Map3k9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100872.1 | NP_001094342.1 | 2 | 776 | ||
| XM_006240303.2 | XP_006240365.1 | 2 | 999 | |||