siRNA Details
| Gene Symbol: Ccnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109672.1 | NP_001103142.1 | 6 | 567 | ||
| XM_006240549.2 | XP_006240611.1 | 7 | 810 | |||
| XM_006240551.2 | XP_006240613.1 | 5 | 812 | |||
| GenBank mRNA | BC129121.1 | - | 6 | 567 | 7125896 | |

Ccnk
Genomic Map