siRNA Details
| Gene Symbol: SLC22A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004256.3 | NP_004247.2 | 1 | 219 | ||
| XM_017007519.1 | XP_016863008.1 | 1 | 120 | |||
| GenBank mRNA | AB010438.1 | - | 1 | 186 | ||
| AK298375.1 | - | 1 | 120 | |||
| AK315301.1 | - | 1 | 214 | |||
| BC035973.1 | - | 1 | 214 | 4609915 | ||
| DA629093.1 | - | 1 | 214 | |||
| DA629311.1 | - | 1 | 219 | |||

SLC22A13
Genomic Map