siRNA Details
| Gene Symbol: Tmem198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109281.1 | NP_001102751.1 | 5 | 1471 | ||
| XM_006240794.2 | XP_006240856.1 | 5 | 1332 | |||
| XM_006240795.1 | XP_006240857.1 | 4 | 1386 | |||
| XM_008765038.1 | XP_008763260.1 | 4 | 1776 | |||

Tmem198b
Genomic Map