siRNA Details
| Gene Symbol: Btbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241037.2 | XP_006241099.2 | 5 | 786 | ||
| XM_008765152.1 | XP_008763374.1 | 5 | 786 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd2| Gene Symbol: Btbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241037.2 | XP_006241099.2 | 5 | 786 | ||
| XM_008765152.1 | XP_008763374.1 | 5 | 786 | |||