siRNA Details
| Gene Symbol: Ccdc105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025774.1 | NP_001020945.1 | 4 | 1054 | ||
| GenBank mRNA | BC098035.1 | - | 4 | 1054 | 7456481 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc105| Gene Symbol: Ccdc105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025774.1 | NP_001020945.1 | 4 | 1054 | ||
| GenBank mRNA | BC098035.1 | - | 4 | 1054 | 7456481 | |