siRNA Details
| Gene Symbol: Slc5a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191987.1 | NP_001178916.1 | 14 | 1685 | ||
| XM_006241232.2 | XP_006241294.1 | 15 | 3460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a8| Gene Symbol: Slc5a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191987.1 | NP_001178916.1 | 14 | 1685 | ||
| XM_006241232.2 | XP_006241294.1 | 15 | 3460 | |||