siRNA Details
| Gene Symbol: Ccer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025052.1 | NP_001020223.1 | 1 | 1381 | ||
| GenBank mRNA | BC079199.1 | - | 1 | 1381 | 7131579 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccer1| Gene Symbol: Ccer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025052.1 | NP_001020223.1 | 1 | 1381 | ||
| GenBank mRNA | BC079199.1 | - | 1 | 1381 | 7131579 | |