siRNA Details
| Gene Symbol: HS6ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004807.2 | NP_004798.3 | 1, 2 | 565 | ||
| GenBank mRNA | AB006179.1 | - | 1, 2 | 639 | ||
| AK293724.1 | - | 1, 2 | 22 | |||
| BC096239.1 | - | 1, 2 | 541 | 40005544 | ||
| BC096240.1 | - | 1, 2 | 541 | 40005547 | ||
| BC099638.3 | - | 1, 2 | 541 | 40005546 | ||
| BC099639.1 | - | 1, 2 | 541 | 40005548 | ||
| BP219645.1 | - | 1, 2 | 451 | |||
| CV569969.1 | - | 2, 3 | 270 | |||
| DR005030.1 | - | 1, 2 | 571 | |||

HS6ST1
Genomic Map