siRNA Details
| Gene Symbol: LOC500846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047962.1 | NP_001041427.1 | 7 | 1056 | ||
| GenBank mRNA | AY539897.1 | - | 7 | 1056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500846| Gene Symbol: LOC500846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047962.1 | NP_001041427.1 | 7 | 1056 | ||
| GenBank mRNA | AY539897.1 | - | 7 | 1056 | ||