siRNA Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130575.1 | NP_001124047.1 | 3 | 1353 | ||
| XM_008765454.1 | XP_008763676.1 | 3 | 1503 | |||
| XM_008765455.1 | XP_008763677.1 | 3 | 802 | |||
| GenBank mRNA | AB232173.1 | - | 3 | 1505 | ||

Rspo2
Genomic Map