siRNA Details
| Gene Symbol: LOC500876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145141.1 | NP_001138613.1 | 1 | 1457 | ||
| GenBank mRNA | BC105880.1 | - | 1 | 1457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500876| Gene Symbol: LOC500876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145141.1 | NP_001138613.1 | 1 | 1457 | ||
| GenBank mRNA | BC105880.1 | - | 1 | 1457 | ||